siRNA Details
| Gene Symbol: Ghsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177330.4 | NP_796304.1 | 3 | 1092 | ||
| GenBank mRNA | AK049671.1 | - | 3 | 1093 | ||
| BC120672.1 | - | 2 | 895 | 40129595 | ||
| BC137882.1 | - | 2 | 895 | 8860901 | ||
More Information
Set Membership: |
|

Ghsr
Genomic Map