siRNA Details
| Gene Symbol: Alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145362.2 | NP_663337.2 | 12 | 1251 | ||
| GenBank mRNA | AK145871.1 | - | 12 | 1251 | ||
| AK185215.1 | - | 329 | ||||
| AK209849.1 | - | 2 | 145 | |||
| BC011281.1 | - | 11 | 1158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg1| Gene Symbol: Alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145362.2 | NP_663337.2 | 12 | 1251 | ||
| GenBank mRNA | AK145871.1 | - | 12 | 1251 | ||
| AK185215.1 | - | 329 | ||||
| AK209849.1 | - | 2 | 145 | |||
| BC011281.1 | - | 11 | 1158 | |||