siRNA Details
| Gene Symbol: Tmem132c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175432.3 | NP_780641.2 | 2 | 1062 | ||
| GenBank mRNA | AK028289.1 | - | 2 | 1081 | ||
| AK028541.1 | - | 2 | 1063 | |||
| AK157823.1 | - | 2 | 1063 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem132c| Gene Symbol: Tmem132c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175432.3 | NP_780641.2 | 2 | 1062 | ||
| GenBank mRNA | AK028289.1 | - | 2 | 1081 | ||
| AK028541.1 | - | 2 | 1063 | |||
| AK157823.1 | - | 2 | 1063 | |||