siRNA Details
| Gene Symbol: Mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172457.2 | NP_766045.2 | 3 | 671 | ||
| GenBank mRNA | AK030481.1 | - | 3 | 707 | ||
| AK170394.1 | - | 3 | 682 | |||
| AK171197.1 | - | 695 | ||||
| BC051170.1 | - | 2 | 660 | |||
| BC058238.1 | - | 3 | 671 | 6307543 | ||

Mob3a
Genomic Map