siRNA Details
| Gene Symbol: Zfp871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172458.3 | NP_766046.3 | 4 | 1087 | ||
| GenBank mRNA | AK029041.1 | - | 4 | 1075 | ||
| AK122579.1 | - | 4 | 1050 | |||
| BC043671.1 | - | 4 | 1202 | 5040597 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp871| Gene Symbol: Zfp871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172458.3 | NP_766046.3 | 4 | 1087 | ||
| GenBank mRNA | AK029041.1 | - | 4 | 1075 | ||
| AK122579.1 | - | 4 | 1050 | |||
| BC043671.1 | - | 4 | 1202 | 5040597 | ||