siRNA Details
| Gene Symbol: Rpp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145938.4 | NP_666050.3 | 4 | 386 | ||
| GenBank mRNA | AK048810.1 | - | 386 | |||
| BC024607.1 | - | 4 | 331 | 4236826 | ||
| BC037666.1 | - | 3 | 1948 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpp40| Gene Symbol: Rpp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145938.4 | NP_666050.3 | 4 | 386 | ||
| GenBank mRNA | AK048810.1 | - | 386 | |||
| BC024607.1 | - | 4 | 331 | 4236826 | ||
| BC037666.1 | - | 3 | 1948 | |||