siRNA Details
| Gene Symbol: Shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040459.1 | NP_001035549.1 | 6 | 3011 | ||
| GenBank mRNA | AK052531.1 | - | 5 | 3054 | ||
| AK173129.1 | - | 5 | 3033 | |||
| BC151022.1 | - | 6 | 3302 | 9087935 | ||
| DQ435686.1 | - | 6 | 3011 | |||
| DQ435687.1 | - | 6 | 3011 | |||

Shroom4
Genomic Map