siRNA Details
| Gene Symbol: Dip2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081426.2 | NP_001074895.2 | 16, 17 | 2332 | ||
| GenBank mRNA | AK173066.1 | - | 8, 9 | 744 | ||
| BC137787.1 | - | 16, 17 | 1924 | 8860806 | ||
| BC144910.1 | - | 16, 17 | 1924 | 9053458 | ||

Dip2c
Genomic Map