siRNA Details
| Gene Symbol: Dip2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081426.2 | NP_001074895.2 | 5 | 973 | ||
| GenBank mRNA | BC137787.1 | - | 5 | 565 | 8860806 | |
| BC144909.1 | - | 6 | 733 | 9053457 | ||
| BC144910.1 | - | 5 | 565 | 9053458 | ||

Dip2c
Genomic Map