siRNA Details
| Gene Symbol: Nek11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172461.3 | NP_766049.2 | 15 | 1828 | ||
| GenBank mRNA | AK030042.1 | - | 15 | 1787 | ||
| BC119588.1 | - | 14 | 1662 | 40058889 | ||
| BC119589.1 | - | 14 | 1662 | 40058890 | ||

Nek11
Genomic Map