siRNA Details
| Gene Symbol: Nek11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172461.3 | NP_766049.2 | 7 | 1001 | ||
| GenBank mRNA | AK030042.1 | - | 7 | 960 | ||
| AK030186.1 | - | 6 | 877 | |||
| AK043543.1 | - | 7 | 1240 | |||
| AK054237.1 | - | 7 | 895 | |||
| BC119588.1 | - | 6 | 835 | 40058889 | ||
| BC119589.1 | - | 6 | 835 | 40058890 | ||

Nek11
Genomic Map