siRNA Details
| Gene Symbol: Mterf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042670.1 | NP_001036135.2 | 2 | 787 | ||
| GenBank mRNA | AK016498.1 | - | 2 | 787 | ||
| BC141337.1 | - | 2 | 758 | 9055949 | ||
| Gene Symbol: Mterf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158221.1 | - | 2 | 1825 | ||
| BC082778.1 | - | 2 | 769 | 3994365 | ||
| BC094442.1 | - | 2 | 778 | 3587753 | ||

Mterf1b
Genomic Map