siRNA Details
| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145939.2 | NP_666051.2 | 4 | 532 | ||
| GenBank mRNA | AK090067.1 | - | 3 | 433 | ||
| AK149510.1 | - | 3 | 431 | |||
| BC031893.1 | - | 4 | 527 | 5371807 | ||
| CX217502.1 | - | 4 | 479 | |||

Alg3
Genomic Map