siRNA Details
| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145939.2 | NP_666051.2 | 6 | 801 | ||
| GenBank mRNA | AK090067.1 | - | 5 | 702 | ||
| AK149510.1 | - | 5 | 701 | |||
| BC031893.1 | - | 6 | 796 | 5371807 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg3| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145939.2 | NP_666051.2 | 6 | 801 | ||
| GenBank mRNA | AK090067.1 | - | 5 | 702 | ||
| AK149510.1 | - | 5 | 701 | |||
| BC031893.1 | - | 6 | 796 | 5371807 | ||