siRNA Details
| Gene Symbol: Slc25a38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144793.1 | NP_659042.1 | 5 | 697 | ||
| GenBank mRNA | AK078385.1 | - | 5 | 713 | ||
| BC010801.1 | - | 5 | 697 | 4235245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a38| Gene Symbol: Slc25a38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144793.1 | NP_659042.1 | 5 | 697 | ||
| GenBank mRNA | AK078385.1 | - | 5 | 713 | ||
| BC010801.1 | - | 5 | 697 | 4235245 | ||