siRNA Details
| Gene Symbol: Creb3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178661.4 | NP_848776.2 | 2 | 624 | ||
| GenBank mRNA | AK035695.1 | - | 2 | 614 | ||
| AK036712.1 | - | 2 | 616 | |||
| AK041050.1 | - | 2 | 597 | |||
| AK041669.1 | - | 2 | 624 | |||
| AK042418.1 | - | 2 | 618 | |||
| BC043466.1 | - | 2 | 586 | 5052843 | ||
| BC138470.1 | - | 2 | 470 | 8861490 | ||
| BC138471.1 | - | 2 | 470 | 8861491 | ||

Creb3l2
Genomic Map