siRNA Details
| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153782.1 | NP_722477.1 | 2 | 1098 | ||
| GenBank mRNA | AK142932.1 | - | 2 | 1122 | ||
| BC011274.1 | - | 2 | 1004 | |||
| BC029169.1 | - | 2 | 1098 | 4947019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam20a| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153782.1 | NP_722477.1 | 2 | 1098 | ||
| GenBank mRNA | AK142932.1 | - | 2 | 1122 | ||
| BC011274.1 | - | 2 | 1004 | |||
| BC029169.1 | - | 2 | 1098 | 4947019 | ||