siRNA Details
| Gene Symbol: Hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291439.1 | NP_001278368.1 | 2 | 536 | ||
| NM_145942.5 | NP_666054.2 | 3 | 595 | |||
| GenBank mRNA | AK031297.1 | - | 3 | 484 | ||
| AK044835.1 | - | 3 | 485 | |||
| AK045094.1 | - | 2 | 424 | |||
| AK078743.1 | - | 2 | 426 | |||
| AK135551.1 | - | 2 | 424 | |||
| AK136171.1 | - | 3 | 484 | |||
| AK144384.1 | - | 2 | 424 | |||
| AK145424.1 | - | 2 | 424 | |||
| AK146042.1 | - | 2 | 426 | |||
| BC023851.1 | - | 3 | 511 | 5347038 | ||
| BC029693.1 | - | 2 | 441 | 5375374 | ||
| BC031363.1 | - | 2 | 443 | 5357048 | ||
| BC034317.1 | - | 1 | 387 | 5366786 | ||

Hmgcs1
Genomic Map