siRNA Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172157.1 | NP_001165628.1 | 14 | 1698 | ||
| NM_153530.2 | NP_705758.1 | 13 | 1656 | |||
| GenBank mRNA | AK029652.1 | - | 14 | 1712 | ||
| AK031180.1 | - | 14 | 1681 | |||
| AK183644.1 | - | 1 | 52 | |||
| BC036177.1 | - | 13 | 1617 | 5004341 | ||

Dis3l2
Genomic Map