siRNA Details
| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 14, 15 | 1462 | ||
| GenBank mRNA | AK085572.1 | - | 12, 13 | 1312 | ||
| CN458387.1 | - | 3, 4 | 387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fanci| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 14, 15 | 1462 | ||
| GenBank mRNA | AK085572.1 | - | 12, 13 | 1312 | ||
| CN458387.1 | - | 3, 4 | 387 | |||