siRNA Details
| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 24 | 2582 | ||
| GenBank mRNA | AK085572.1 | - | 22 | 2432 | ||
| BC027836.1 | - | 22 | 2309 | |||
| CO426844.1 | - | 2 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fanci| Gene Symbol: Fanci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145946.2 | NP_666058.2 | 24 | 2582 | ||
| GenBank mRNA | AK085572.1 | - | 22 | 2432 | ||
| BC027836.1 | - | 22 | 2309 | |||
| CO426844.1 | - | 2 | 139 | |||