siRNA Details
| Gene Symbol: Dock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153413.2 | NP_700462.2 | 15 | 1670 | ||
| GenBank mRNA | AY145302.1 | - | 15 | 1620 | ||
| CK781165.1 | - | 4 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock3| Gene Symbol: Dock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153413.2 | NP_700462.2 | 15 | 1670 | ||
| GenBank mRNA | AY145302.1 | - | 15 | 1620 | ||
| CK781165.1 | - | 4 | 359 | |||