siRNA Details
| Gene Symbol: Slc26a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145947.2 | NP_666059.2 | 12 | 1526 | ||
| GenBank mRNA | AF345194.1 | - | 11 | 1428 | ||
| AK054327.1 | - | 12 | 1527 | |||
| BC026928.1 | - | 12 | 1505 | 4020760 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a7| Gene Symbol: Slc26a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145947.2 | NP_666059.2 | 12 | 1526 | ||
| GenBank mRNA | AF345194.1 | - | 11 | 1428 | ||
| AK054327.1 | - | 12 | 1527 | |||
| BC026928.1 | - | 12 | 1505 | 4020760 | ||