siRNA Details
| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081153.1 | NP_001074622.1 | 27, 28 | 5873 | ||
| GenBank mRNA | AK134584.1 | - | 1, 2 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13c| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081153.1 | NP_001074622.1 | 27, 28 | 5873 | ||
| GenBank mRNA | AK134584.1 | - | 1, 2 | 70 | ||