siRNA Details
| Gene Symbol: Adamts18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172466.2 | NP_766054.2 | 3 | 592 | ||
| GenBank mRNA | AK035797.1 | - | 3 | 492 | ||
| AK048211.1 | - | 1 | 185 | |||
| AK053851.1 | - | 2 | 700 | |||
| AK081122.1 | - | 3 | 495 | |||
| AK140876.1 | - | 3 | 368 | |||
| BC094674.1 | - | 3 | 247 | 30548647 | ||
| CB246179.1 | - | 2 | 241 | |||

Adamts18
Genomic Map