siRNA Details
| Gene Symbol: Thnsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001297.2 | NP_001001297.1 | 3 | 417 | ||
| NM_177588.2 | NP_808256.1 | 3 | 1989 | |||
| GenBank mRNA | AK029518.1 | - | 3 | 1990 | ||
| AK033465.1 | - | 3 | 386 | |||
| AK083895.1 | - | 3 | 1989 | |||
| AK087148.1 | - | 3 | 499 | |||
| BC027743.1 | - | 1 | 514 | |||
| BC090667.1 | - | 1 | 1698 | 30686917 | ||
| BC117854.2 | - | 2 | 1773 | 40094242 | ||

Thnsl1
Genomic Map