siRNA Details
| Gene Symbol: Thnsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001297.2 | NP_001001297.1 | 3 | 570 | ||
| NM_177588.2 | NP_808256.1 | 3 | 2142 | |||
| GenBank mRNA | AK029518.1 | - | 3 | 2143 | ||
| AK033465.1 | - | 3 | 539 | |||
| AK083895.1 | - | 3 | 2142 | |||
| BC027743.1 | - | 1 | 667 | |||
| BC090667.1 | - | 1 | 1851 | 30686917 | ||
| BC117854.2 | - | 2 | 1926 | 40094242 | ||

Thnsl1
Genomic Map