siRNA Details
| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166648.1 | NP_001160120.1 | 8 | 816 | ||
| NM_001166649.1 | NP_001160121.1 | 8 | 837 | |||
| NM_001166650.1 | NP_001160122.1 | 7 | 775 | |||
| NM_153532.3 | NP_705760.2 | 7 | 1147 | |||
| GenBank mRNA | AK129399.1 | - | 8 | 796 | ||
| AK141491.1 | - | 8 | 817 | |||
| BC028839.1 | - | 7 | 787 | 5150255 | ||
| BC051397.1 | - | 7 | 775 | 5150255 | ||
| BC063775.1 | - | 7 | 1159 | 5375514 | ||

Zfp280c
Genomic Map