siRNA Details
| Gene Symbol: Fam83b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045518.1 | NP_001038983.1 | 4 | 1419 | ||
| GenBank mRNA | AK049628.1 | - | 5 | 1511 | ||
| AK145108.1 | - | 5 | 1504 | |||
| BC120577.1 | - | 4 | 1419 | 40129500 | ||
| BC120579.1 | - | 4 | 1419 | 40129502 | ||

Fam83b
Genomic Map