siRNA Details
| Gene Symbol: Snx30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172468.2 | NP_766056.1 | 7 | 1466 | ||
| GenBank mRNA | AK029012.1 | - | 7 | 1268 | ||
| AK076374.1 | - | 4 | 470 | |||
| AK147437.1 | - | 7 | 1468 | |||
| AK165619.1 | - | 7 | 1276 | |||
| AK170774.1 | - | 7 | 1240 | |||
| AK170999.1 | - | 7 | 1268 | |||
| BC099674.1 | - | 7 | 1262 | 4950374 | ||

Snx30
Genomic Map