siRNA Details
| Gene Symbol: Wfdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145369.3 | NP_663344.2 | 4 | 1213 | ||
| GenBank mRNA | AK131905.1 | - | 4 | 1161 | ||
| AY669505.1 | - | 4 | 1212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc5| Gene Symbol: Wfdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145369.3 | NP_663344.2 | 4 | 1213 | ||
| GenBank mRNA | AK131905.1 | - | 4 | 1161 | ||
| AY669505.1 | - | 4 | 1212 | |||