siRNA Details
| Gene Symbol: Csta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033239.3 | NP_001028411.1 | 3 | 380 | ||
| GenBank mRNA | AK137702.1 | - | 3 | 381 | ||
| BC139156.1 | - | 3 | 275 | 8862178 | ||
| BC139157.1 | - | 3 | 275 | 8862179 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csta1| Gene Symbol: Csta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033239.3 | NP_001028411.1 | 3 | 380 | ||
| GenBank mRNA | AK137702.1 | - | 3 | 381 | ||
| BC139156.1 | - | 3 | 275 | 8862178 | ||
| BC139157.1 | - | 3 | 275 | 8862179 | ||