siRNA Details
| Gene Symbol: Gen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177331.4 | NP_796305.3 | 8 | 1045 | ||
| GenBank mRNA | AK030995.1 | - | 8 | 1055 | ||
| AK031077.1 | - | 8 | 1024 | |||
| AK158255.1 | - | 7 | 868 | |||
| BC090653.1 | - | 8 | 1045 | 30659763 | ||
| BC099936.1 | - | 7 | 854 | |||
| BC138157.1 | - | 8 | 932 | 8861176 | ||
| BC138158.1 | - | 8 | 932 | 8861177 | ||

Gen1
Genomic Map