siRNA Details
| Gene Symbol: Trim30d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167828.1 | NP_001161300.1 | 5 | 621 | ||
| NM_199146.2 | NP_954597.2 | 6 | 1097 | |||
| GenBank mRNA | AK136456.1 | - | 5 | 625 | ||
| AK164938.1 | - | 5 | 620 | |||
| BC056631.1 | - | 6 | 1073 | 5035734 | ||

Trim30d
Genomic Map