siRNA Details
| Gene Symbol: Tmem164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199357.1 | NP_001186286.1 | 2, 3 | 200 | ||
| NM_001199360.1 | NP_001186289.1 | 3, 4 | 916 | |||
| NM_177592.4 | NP_808260.2 | 3, 4 | 859 | |||
| GenBank mRNA | AK089342.1 | - | 3, 4 | 766 | ||
| AK165839.1 | - | 3, 4 | 752 | |||
| AK165910.1 | - | 3, 4 | 752 | |||
| AK166488.1 | - | 2, 3 | 186 | |||
| AK199343.1 | - | 2, 3 | 66 | |||
| BC056470.1 | - | 3, 4 | 841 | 5706838 | ||

Tmem164
Genomic Map