siRNA Details
| Gene Symbol: Taar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007266.1 | NP_001007267.1 | 2 | 42 | ||
| GenBank mRNA | AY702326.1 | - | 2 | 42 | ||
| BC139118.1 | - | 2 | 95 | 8862140 | ||
| BC145814.1 | - | 2 | 95 | 40131117 | ||
More Information
Set Membership: |
|

Taar2
Genomic Map