siRNA Details
| Gene Symbol: Tyw3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168358.1 | NP_001161830.1 | 5 | 588 | ||
| NM_172474.5 | NP_766062.1 | 6 | 722 | |||
| GenBank mRNA | AK030344.1 | - | 6 | 720 | ||
| AK034820.1 | - | 6 | 707 | |||
| BC086686.1 | - | 6 | 696 | 30680486 | ||
| BC087879.1 | - | 6 | 686 | 6533208 | ||
| BC141040.1 | - | 6 | 701 | 9055652 | ||

Tyw3
Genomic Map