siRNA Details
Gene Symbol: Sectm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145373.2 | NP_663348.2 | 5 | 659 | ||
GenBank mRNA | AK082020.1 | - | 4 | 351 | ||
BC010462.1 | - | 5 | 626 | 3585040 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sectm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145373.2 | NP_663348.2 | 5 | 659 | ||
GenBank mRNA | AK082020.1 | - | 4 | 351 | ||
BC010462.1 | - | 5 | 626 | 3585040 |