siRNA Details
| Gene Symbol: Bend7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190400.1 | NP_001177329.1 | 6 | 1439 | ||
| NM_178663.4 | NP_848778.1 | 6 | 1439 | |||
| GenBank mRNA | AK030428.1 | - | 6 | 1126 | ||
| AK042822.1 | - | 6 | 1437 | |||
| AK053896.1 | - | 6 | 1441 | |||
| BC056476.1 | - | 3 | 483 | |||
| BC096430.1 | - | 6 | 1111 | 5041388 | ||
| BC119370.1 | - | 6 | 1078 | 8734119 | ||
| BC144864.1 | - | 6 | 1078 | 9053412 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191834.

Bend7
Genomic Map