siRNA Details
Gene Symbol: Dhtkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081131.2 | NP_001074600.1 | 3 | 608 | ||
GenBank mRNA | AK049272.1 | - | 599 | |||
BC117994.1 | - | 559 | 40097991 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhtkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081131.2 | NP_001074600.1 | 3 | 608 | ||
GenBank mRNA | AK049272.1 | - | 599 | |||
BC117994.1 | - | 559 | 40097991 |