siRNA Details
| Gene Symbol: Lcorl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163073.1 | NP_001156545.1 | 5 | 569 | ||
| NM_172153.3 | NP_742165.1 | 5 | 582 | |||
| NM_178142.5 | NP_835278.2 | 5 | 569 | |||
| GenBank mRNA | AB076078.1 | - | 5 | 581 | ||
| AK041987.1 | - | 5 | 528 | |||
| AK042567.1 | - | 5 | 583 | |||
| AK043498.1 | - | 5 | 527 | |||
| AK043845.1 | - | 5 | 537 | |||
| AK154398.1 | - | 5 | 530 | |||
| AK155421.1 | - | 5 | 530 | |||
| BC066151.1 | - | 5 | 499 | |||
| BC138563.1 | - | 5 | 569 | 8861583 | ||

Lcorl
Genomic Map