siRNA Details
| Gene Symbol: Tmc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172476.4 | NP_766064.2 | 14, 15 | 2048 | ||
| GenBank mRNA | AK083189.1 | - | 14, 15 | 2051 | ||
| AK132820.1 | - | 14, 15 | 2080 | |||
| AY236499.1 | - | 14, 15 | 2109 | |||
| AY263161.1 | - | 14, 15 | 2077 | |||
| BC105966.1 | - | 8, 9 | 1062 | |||

Tmc7
Genomic Map