siRNA Details
| Gene Symbol: Tmc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172476.4 | NP_766064.2 | 14 | 1995 | ||
| GenBank mRNA | AK083189.1 | - | 14 | 1998 | ||
| AK132820.1 | - | 14 | 2027 | |||
| AY236499.1 | - | 14 | 2056 | |||
| AY263161.1 | - | 14 | 2024 | |||
| BC105966.1 | - | 8 | 1009 | |||

Tmc7
Genomic Map