siRNA Details
| Gene Symbol: Gpr139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024138.1 | NP_001019309.1 | 2 | 909 | ||
| GenBank mRNA | AB196532.1 | - | 2 | 909 | ||
| AY485344.1 | - | 2 | 909 | |||
| BC125496.1 | - | 2 | 983 | 40130011 | ||
| BC125498.1 | - | 2 | 983 | 40130013 | ||

Gpr139
Genomic Map