siRNA Details
| Gene Symbol: Tbc1d25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172478.3 | NP_766066.2 | 1 | 288 | ||
| GenBank mRNA | AK040932.1 | - | 1 | 31 | ||
| AK078145.1 | - | 1 | 289 | |||
| BC060178.1 | - | 1 | 65 | |||
| BC128566.1 | - | 1 | 238 | 40094900 | ||

Tbc1d25
Genomic Map