siRNA Details
| Gene Symbol: Pgbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171824.2 | NP_741958.1 | 4 | 858 | ||
| GenBank mRNA | BC094384.1 | - | 856 | 30606749 | ||
| CA316156.1 | - | 3 | 661 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgbd5| Gene Symbol: Pgbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171824.2 | NP_741958.1 | 4 | 858 | ||
| GenBank mRNA | BC094384.1 | - | 856 | 30606749 | ||
| CA316156.1 | - | 3 | 661 | |||