siRNA Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178664.5 | NP_848779.2 | 6 | 569 | ||
| GenBank mRNA | AK031361.1 | - | 6 | 558 | ||
| AK043722.1 | - | 6 | 572 | |||
| AK045461.1 | - | 5 | 426 | |||
| AK156318.1 | - | 6 | 547 | |||
| BC088993.1 | - | 6 | 434 | 6416623 | ||
| BC125626.1 | - | 6 | 542 | 40130141 | ||
| BC132193.1 | - | 6 | 542 | 40130470 | ||
| BC145163.1 | - | 6 | 546 | |||
| BC145165.1 | - | 6 | 608 | |||
| BC145166.1 | - | 6 | 542 | 9053717 | ||

B3gntl1
Genomic Map