siRNA Details
| Gene Symbol: Metrnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144797.3 | NP_659046.1 | 4 | 1965 | ||
| GenBank mRNA | AK034855.1 | - | 4 | 1966 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Metrnl| Gene Symbol: Metrnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144797.3 | NP_659046.1 | 4 | 1965 | ||
| GenBank mRNA | AK034855.1 | - | 4 | 1966 | ||