siRNA Details
| Gene Symbol: Nlrp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172481.2 | NP_766069.1 | 3 | 253 | ||
| GenBank mRNA | AK054426.1 | - | 3 | 254 | ||
| AY355342.1 | - | 3 | 254 | |||
| AY596198.1 | - | 1 | 89 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp4b| Gene Symbol: Nlrp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172481.2 | NP_766069.1 | 3 | 253 | ||
| GenBank mRNA | AK054426.1 | - | 3 | 254 | ||
| AY355342.1 | - | 3 | 254 | |||
| AY596198.1 | - | 1 | 89 | |||