siRNA Details
| Gene Symbol: Zfp658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008549.2 | NP_001008549.1 | 5 | 2300 | ||
| GenBank mRNA | AK155078.1 | - | 5 | 2302 | ||
| BC043301.1 | - | 1 | 752 | |||
| BC087557.1 | - | 4 | 2235 | 30742836 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp658| Gene Symbol: Zfp658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008549.2 | NP_001008549.1 | 5 | 2300 | ||
| GenBank mRNA | AK155078.1 | - | 5 | 2302 | ||
| BC043301.1 | - | 1 | 752 | |||
| BC087557.1 | - | 4 | 2235 | 30742836 | ||